Water purification system, RiOs™ 3
Über diesen Artikel
Suitable for general, non-critical laboratory applications or as a pretreatment for ultrapure water systems, the RiOs™ 3 system provides an ideal solution for Type III laboratory grade water needs such as manual cleaning and rinsing, buffer preparation, feed water for humidifiers, autoclaves and glassware washers. Designed for users seeking a compact, easy to use solution, the RiOs™ 3 system will be highly appreciated for its intuitive operation, small footprint and ease of maintenance. For users needing access to ultrapure water, the RiOs™ 3 system is ideal as pretreatment for Synergy® or Milli-Q® water purification systems. Up to 30 L/day of high quality analytical grade water (Type III water) from tap water.
- Easy to install and to use
- Unique all-in-one pack that includes the pretreatment and reverse osmosis membrane
- Maintenance is reduced to the change of a single cartridge once or twice per year
- Backlit colour display shows conductivity, temperature and water level in the integrated reservoir
- Integrated, compact 6 litre reservoir – connection to a 30 or 60 l reservoir is possible for higher daily needs
Bestellinformationen: The purification pack SmartPak™ (171-0126) has to be ordered with the system.
1 mögliche Produktvariationen vorhanden
Product Details & Documents
Suitable for general, non-critical laboratory applications or as a pretreatment for ultrapure water systems, the RiOs™ 3 system provides an ideal solution for Type III laboratory grade water needs such as manual cleaning and rinsing, buffer preparation, feed water for humidifiers, autoclaves and glassware washers. Designed for users seeking a compact, easy to use solution, the RiOs™ 3 system will be highly appreciated for its intuitive operation, small footprint and ease of maintenance. For users needing access to ultrapure water, the RiOs™ 3 system is ideal as pretreatment for Synergy® or Milli-Q® water purification systems. Up to 30 L/day of high quality analytical grade water (Type III water) from tap water.
- Easy to install and to use
- Unique all-in-one pack that includes the pretreatment and reverse osmosis membrane
- Maintenance is reduced to the change of a single cartridge once or twice per year
- Backlit colour display shows conductivity, temperature and water level in the integrated reservoir
- Integrated, compact 6 litre reservoir – connection to a 30 or 60 l reservoir is possible for higher daily needs
Bestellinformationen: The purification pack SmartPak™ (171-0126) has to be ordered with the system.
Spezifikationen für diese Produktfamilie
Spezifikationen:
- Catalog No:
- 171-0126
- 171-0139
- 171-0316
- 171-0317
- 171-0391
- 171-0142
- 171-0315
- Model:
- RiOs™ 3
- RiOs™ 3
- RiOs™ 3
- RiOs™ 3
- RiOs™ 3
- RiOs™ 3
- Production flow rate:
- 3 L/h @ 15 °C
- 3 L/h @ 15 °C
- 3 L/h @ 15 °C
- 3 L/h @ 15 °C
- 3 L/h @ 15 °C
- 3 L/h @ 15 °C
- Microorganisms:
- 99% (organics, particulates), 96% (ionic content) rejected typically
- 99% (organics, particulates), 96% (ionic content) rejected typically
- 99% (organics, particulates), 96% (ionic content) rejected typically
- 99% (organics, particulates), 96% (ionic content) rejected typically
- 99% (organics, particulates), 96% (ionic content) rejected typically
- 99% (organics, particulates), 96% (ionic content) rejected typically
- W×D×H:
- 290×330×500 mm
- 290×330×500 mm
- 290×330×500 mm
- 290×330×500 mm
- 290×330×500 mm
- 290×330×500 mm
- Weight:
- 7 kg
- 7 kg
- 7 kg
- 7 kg
- 7 kg
- 7 kg
Spezifikationen:
Product Family Options
Cartridges
Produktinformationen
Accessories
Produktinformationen
- Catalog #AvailabilityPrice
- Tank connection kit
Produktinformationen
- Catalog #AvailabilityPrice
- Wall-mounting bracket with tap, for Direct Q® and RiOs™Wall-mounting bracket with tap, for Direct Q® and RiOs™171-0759Anmelden für die Verfügbarkeit1 * 1 ST - 298,00 €
Produktinformationen
- Catalog #AvailabilityPrice
- Vent filter for 30/60/100 l PE reservoirsVent filter for 30/60/100 l PE reservoirs172-5097Anmelden für die Verfügbarkeit1 * 1 ST - 147,00 €
Produktinformationen
- Catalog #AvailabilityPrice
- Vent filter for internal tank, prevents contamination of the pure water stored in 6/10 l internal storage tanksVent filter for internal tank, prevents contamination of the pure water stored in 6/10 l internal storage tanks171-0550Anmelden für die Verfügbarkeit1 * 2 ST - 99,70 €
Produktinformationen
- Catalog #AvailabilityPrice
- 30 l PE pure water storage reservoir
Produktinformationen
- Catalog #AvailabilityPrice
- Pressure regulator
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.



