Orders LinkContactUsLinkComponent
UV transilluminators, multibrand
UV transilluminators, multibrand
Supplier:   Vilber Lourmat
UV transilluminators, multibrand
Supplier:  Vilber Lourmat
Certificates
UV transilluminators, multibrand
UV transilluminators, multibrand
6 Options Available Below
Product Details & Documents
This UV transilluminator accommodates a wide range of applications requiring different wavelengths for both visualisation and documentation.
  • UV protection shield
  • Two UV wavelengths in one single transilluminator
  • Super-Bright filter to stop the visible light from UV tubes for an unbeatable viewing field
15-Watt W×D×H: 470×330×150 mm
Product Family Options
Product Information
  • Model
    Output
    Filter size
    Wavelength
    Availability
    Price
  • TCP-20.LC V1
    5×8 / 6×8 W
    200×200 mm
    365 / 254 nm
    Log in for availability
    Log in for availability
  • TCP-20.MC V1
    5×8 / 6×8 W
    200×200 mm
    312 / 254 nm
    Log in for availability
    Log in for availability
  • TCP-26.LC V1
    5×8 / 6×8 W
    210×260 mm
    365 / 254 nm
    Log in for availability
    Log in for availability
    - 1 700,00 €
    1 700,00 €
       1 700,00 €
  • TCP-26.LM V1
    5×8 / 6×8 W
    210×260 mm
    365 / 312 nm
    Log in for availability
    Log in for availability
    - 1 700,00 €
    1 700,00 €
       1 700,00 €
  • TCP-26.LMX V1
    5×8 / 6×8 W
    210×260 mm
    365 / 312 nm
    Log in for availability
    Log in for availability
    - 2 130,00 €
    2 130,00 €
       2 130,00 €
  • TCP-26.MC V1
    5×8 / 6×8 W
    210×260 mm
    312 / 254 nm
    Log in for availability
    Log in for availability
    - 1 700,00 €
    1 700,00 €
       1 700,00 €