Orders LinkContactUsLinkComponent
Digital timer, three channels
Digital timer, 3 channels, Grey, 65×20×55 mm
  MOIN419CA
 :   MOINEAU
Digital timer, three channels
  MOIN419CA
 :  MOINEAU
Digital timer, three channels
Digital timer, three channels
Digital timer, 3 channels, Grey, 65×20×55 mm
56,40 €
1 * 1 ST
Three-channel (1× count up, 2× count down) timer with metal housing.
  • Timing capacity: 24 hours
  • Two alarms, two memories, snooze function
  • Clock (AM/PM)
 : Supplied with stand, clip, magnet and LR44 battery.
  • Colour:
    Grey
  • Description:
    Digital timer, 3 channels
  • W×D×H:
    65×20×55 mm