KPG stirrer shafts and stirrer bearings
Supplier: witeg Labortechnik
Certificates
About this item
DURAN®, borosilicate glass interchangeable stirrer shafts and bearings.
- WB type stirring shafts with ground and polished bearing surface
- HA, HB, HQ, HT type stirring bearings with ground and polished surface