Order Entry
Germany
Orders LinkContactUsLinkComponent
Magnetic Stirrer, explosion proof

Magnetic Stirrer, explosion proof

Magnetic stirrer, air or water driven
Catalog # 442-4165
Supplier:   KARTELL
Magnetic Stirrer, explosion proof
Catalog # 442-4165
Supplier:  KARTELL
Magnetic Stirrer, explosion proof
Magnetic Stirrer, explosion proof

Magnetic stirrer, air or water driven

Price
178,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Magnetic stirrer with ABS housing that has no electric motor but is powered by either air (1 to 2,5 kg/cm²; 15 to 40 psi) or water pressure.

  • Ideal for environments where flammable solvents and acid vapours are in high concentration
  • No electric motor so risk of sparking hazard is removed
  • Corrosion proof plastic construction
  • Can be used on the bench or clamper to a 12 mm Ø stand

Ordering information: Retort rods Ø 12 mm and stand to be ordered separately

Specifications
  • Description:
    Magnetic stirrer, air or water driven
  • Plate material:
    ABS
  • Cat. No.:
    442-4165
  • Speed range:
    0 - 800 min⁻¹
  • Plate dimensions:
    128 mm
  • Max. stirring capacity H₂O:
    1 L
  • Overall dimensions:
    Ø×H: 129×61,3
  • Weight:
    0,26 kg