Orders LinkContactUsLinkComponent
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse
Supplier:   BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Supplier:  BELLINGHAM STANLEY
Certificates
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse
4 Options Available Below
Product Details & Documents
Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.
  • Shaded field of view for distinction
  • Anti-roll support and ergonomic design
  • Comfortable eyepiece with smooth adjustment
  • Easy to clean prism and push-on flap protection
  • Unique sample ‘dribble’ feature
Delivery information: Supplied with a soft carrying case and instructions.
Product Family Options
Product Information
  • Range
    Resolution
    Availability
    Price
  • 635-0037
    Handheld refractometer for sugar
    Handheld refractometer for sugar
    0 - 15 % Bx
    0,1 % Bx
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • 635-0038
    Handheld refractometer for sugar
    Handheld refractometer for sugar
    0 - 30 % Bx
    0,2 % Bx
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • 635-0048
    Handheld refractometer for starch
    Handheld refractometer for starch
    0 − 30 %
    0,2 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 353,00 €
    353,00 €
    1 * 1 ST
       353,00 €
ERSA
Product Information
  • Range
    Resolution
    Availability
    Price
  • BELL45-006
    Eclipse spare illuminator flaps
    Eclipse spare illuminator flaps
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 55,50 €
    55,50 €
    1 * 3 ST
       55,50 €