Order Entry
Germany
Orders LinkContactUsLinkComponent
Pump, piston siphon

Pump, piston siphon

Piston siphon pump with PE suction tube (590 mm), for containers up to 60 litres, 10 l/min
Catalog # COLL5900
Supplier:   NRG DEVELOPMENTS
Pump, piston siphon
Catalog # COLL5900
Supplier:  NRG DEVELOPMENTS
Pump, piston siphon
Pump, piston siphon

Piston siphon pump with PE suction tube (590 mm), for containers up to 60 litres, 10 l/min

Price
65,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Piston operated siphon pump. Adjustable bung allows the pump to be used with different containers.

  • With PE suction tube
  • For drums up to 205 litres
Specifications
  • Flow rate:
    10 l/min
  • Description:
    Piston siphon pump with PE suction tube (590 mm), for containers up to 60 litres
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Adapter tri-sure for 205 litre pump
    Adapter tri-sure for 205 litre pump
    181-0300
    Log in for availability
    1 * 1 ST - 20,90 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.