Order Entry
Germany
ContactUsLinkComponent
 
Drum pump, gas-tight
  223-1412
 :  BURKLE
undefined
Drum pump, gas-tight
  223-1412
 :  BURKLE

 

  • Flow rate:
    200 ml/Hub
  • Description:
    Drum pump, gas-tight with universal adapter
  • Material:
    PP/PVC
  • Pk:
    1

 

 

These drum pumps prevent the escape of dangerous or hazardous vapours when decanting liquids. The vapours produced are fed back to the decanting container from the filling container via the gas displacement line. The gas and liquid lines are also equipped with stop valves.

  • With universal adapter for container openings from 34 to 70 mm
  • Stepless adjustable telescopic pipe for drum depths of 400 to 670 mm

PP contact parts and flexible PVC tubing.