Digital hotplates
Supplier: NICKEL ELECTRO
Certificates
About this item
Digital hotplates with a cast aluminium plate. Precision electronic controller provides increased sensitivity. Stainless steel casing, stove enamelled finish with fitting for optional retort rod.
- Allow user selectable time and temperature parameters with memory
- Membrane touch control panel and bright display
- Fitted with non-slip rubber feet for safety
Timer settings: 0 to 9999 min.
Temperature uniformity: ±0,5 °C at 60 °C.