Laboratory glassware washer, G 7893
: MIELE
These front loading devices in stainless steel can fit under a work surface and have a large washing compartment with two levels. They are suitable for washing, to analytically pure standards, in industry, environmental and research laboratories. The machines are fitted with an integrated hot air drier and are only 600 mm wide. With integrated hot air dryer.
- Direct coupling of the upper basket, either with spray arm or injector jets, to the water supply enables the items to be thoroughly cleaned inside and out
- Drying Plus enables the machine and also complex items to be effectively dried inside and out
- Integrated dosage pump for an accurate and automatic dosage of neutralisation agents
- Rinsing with demineralised water for analytically pure treatment results
- Profi-Monobloc water softener ensures continuous regeneration during cleaning
- Steam condenser prevents expelled moisture and hot air from damaging room; no connection to extractor required
Compliant with VDE, EMC/radio interference protection, IP X1
: Compliant with VDE, EMC/radio interference protection, IP X1.
: Basket and accessories must be ordered separately. For use as a standalone device, the cover must be ordered separately, please enquire for details.