Orders LinkContactUsLinkComponent
Gel analysis software, Bio-1D
Bio-1D software, 1 licence
Catalog # VILB141010011
Supplier:   Vilber Lourmat
Gel analysis software, Bio-1D
Catalog # VILB141010011
Supplier:  Vilber Lourmat
Gel analysis software, Bio-1D
Gel analysis software, Bio-1D
Bio-1D software, 1 licence
Price
1 730,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Bio-1D is sophisticated and intuitive analysis software designed for use with fluorescent or chemiluminescent samples. The software works with DNA, RNA, proteins, polynucleotides, petri dishes, microplates, plants, and in vivo images.
  • Transforms 1-D gels into 3-D results
  • Automatically detects lanes and bands with the one click Detector™ feature
  • Analysis can be saved as a template and re-used to facilitate routine analysis
  • State-of-the-art analysis algorithms
  • Software modules available include molecular weight calculation, volume quantification, array analysis and image enhancement
Specifications
  • Description:
    Bio-1D software, 1 licence
  • Seq:
    01