Gas cylinder holder
159-9110
- Type:Gas cylinder holder
- Pk:1
Powder coated aluminium gas cylinder holder with safety belt and snap buckles. Can be easily fastened onto workbenches, tables, shelves or a ledge up to 70 mm thickness.
- Suitable for all cylinders size 10 litres and above
- Secures tightly, can be loosened quickly