Crude fibre digestion systems
Catalog # 705-0472
Supplier: GERHARDT
Specifications
- Pk:1
- Type:HY 16/6
Specifications
About this item
Hydrolysis unit, HY 16/6.
Consists of the crude fibre digestion unit RF 16/6 plus a filter insert with 6 funnels DIN 12445, 100 mm, 1 soft polyethylene bottle with a narrow mouth (1000 ml), 100 folded filters as well as 250 g boiling stones.
Consists of the crude fibre digestion unit RF 16/6 plus a filter insert with 6 funnels DIN 12445, 100 mm, 1 soft polyethylene bottle with a narrow mouth (1000 ml), 100 folded filters as well as 250 g boiling stones.
Delivery: Supplied with special supports, glass condensers, ring shaped rest, silicone rubber tubings, beakers (1000 ml), and power cord.