Joulemeter with cover
Calorimeter
Catalog # 761-0049
Supplier: BREUKHOVEN
Jump to:
Product Details & Documents
For determining the specific heat of solids and liquids. Aluminium interior, plastic exterior. Plastic cover with slot for thermometer, stirring rod, and immersion thermostat of 12 ohms with a 4 mm connector.
- Voltage: 10 V
- Capacity: 200 ml
Height: 130 mm, Diameter: 95 mm
Specifications
- Description:Calorimeter
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



