Bottle-top dispensers, EM-dispenser
Supplier: HIRSCHMANN
Certificates
About this item
PP with glass piston, variable volume, autoclavable.
- For mild, aqueous media or organic liquids that do not attack PP
- Micrometric adjustment knob for fine tuning
- Certificate of quality and conformity
Ordering information: Supplied with A 32 thread, three adapters (A 28, A 45, S 40), an aspiration tube with screw connection, an evacuation tube, a serial number and a quality certificate.