Orders LinkContactUsLinkComponent
VWR®, Speciality pH Electrodes
VWR®, Speciality pH Electrodes
VWR®, Speciality pH Electrodes
Supplier:  VWR Collection
Certificates
VWR®, Speciality pH Electrodes
VWR®, Speciality pH Electrodes
5 Options Available Below
Product Family Options
Product Information
  • Type
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    For
    Availability
    Price
  • Flat extra wide, double junction (HDPE/Pellon)
    +10...+100 °C
    Epoxy
    Gel
    15×115 mm
    BNC
    Agar, microbiological samples, cheese, butter, water
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 169,00 €
    169,00 €
    1 * 1 ST
       169,00 €
  • Low ionic, double juntion (Pellon)
    0...+100 °C
    Epoxy
    Gel
    12×150 mm
    BNC
    Low ionic strength solutions
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  • ORP (Redox), Pellon junction
    0...+100 °C
    Epoxy
    Gel
    12×150 mm
    BNC
    Electroplating baths
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • pH only half cell
    0...+100 °C
    Epoxy
    12×150 mm
    BNC
    Water, creams, beverage
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • pH only half cell
    0...+100 °C
    Glass
    12×150 mm
    DIN
    Water, creams, beverage
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €