Wipe rolls, WYPALL* L40
115-2051
- Packed:1 roll of 750 sheets
- Colour:White
- Description:Large roll, 1 ply, perforated
- L×W:315×340 mm
- Seq:01
- Pk:1 Roll
These single ply wipes are made from AIRFLEX* material, a specialised fabric with a perfect balance of softness and strength, that is also thick and absorbent. Ideal for industrial tasks such as cleaning, process line wiping, soaking up large spills.
- Made with double re-creped (DRC) base sheet technology
- Large, perforated rolls for use in busy areas
- Also suitable for use in healthcare for wiping patients’ hands and face
- Can be used with a dispensing system to help control use and reduce wastage
Matching dispensers: 115-2038, 115-0045 and 115-1533.