Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Supplier:   Kavalier
Watch glasses
Supplier:  Kavalier
Certificates
Watch glasses
5 Options Available Below
Product Details & Documents
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fused edges.
Certifications: DIN 12341
Documents
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • 216-1175
    Watch glass, soda glass, with fused edges
    Watch glass, soda glass, with fused edges
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,30 €
    5,30 €
    1 * 1 ST
       5,30 €
  • 216-1176
    Watch glass, soda glass, with fused edges
    Watch glass, soda glass, with fused edges
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,40 €
    5,40 €
    1 * 1 ST
       5,40 €
  • 216-1177
    Watch glass, soda glass, with fused edges
    Watch glass, soda glass, with fused edges
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,70 €
    5,70 €
    1 * 1 ST
       5,70 €
  • 216-1178
    Watch glass, soda glass, with fused edges
    Watch glass, soda glass, with fused edges
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,80 €
    4,80 €
    1 * 1 ST
       4,80 €
  • 216-1180
    Watch glass, soda glass, with fused edges
    Watch glass, soda glass, with fused edges
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,50 €
    6,50 €
    1 * 1 ST
       6,50 €