Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Supplier:   KARTELL
Watch glasses
Supplier:  KARTELL
Certificates
Watch glasses
4 Options Available Below
Product Details & Documents
PP, transparent.
  • Excellent chemical resistance
  • Heat resistant up to +120 °C
  • Autoclavable
Stable, with extra wide foot.
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • 216-1134
    Watch glass, PP, moulded base
    Watch glass, PP, moulded base
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,20 €
    5,20 €
    1 * 1 ST
       5,20 €
  • 216-1136
    Watch glass, PP, moulded base
    Watch glass, PP, moulded base
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,60 €
    6,60 €
    1 * 1 ST
       6,60 €
  • 216-1138
    Watch glass, PP, moulded base
    Watch glass, PP, moulded base
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,70 €
    7,70 €
    1 * 1 ST
       7,70 €
  • 216-1140
    Watch glass, PP, moulded base
    Watch glass, PP, moulded base
    119 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,50 €
    8,50 €
    1 * 1 ST
       8,50 €