WHEATON® Aluminium Seals, Unlined
New Product
Aluminum seals (unlined) are available in a wide range of styles and sizes.
- Open top seals expose stopper for sample retrieval with a syringe
- Tear-off seal removes completely allowing content to pour from bottle
- Solid top seals are excellent for long-term storage of samples
- Flip cap seals provide tamper evidence
Colour selection includes natural, red, blue, and green.