ContactUsLinkComponent
Guillotine tubing cutter
Guillotine tubing cutter
Catalog # 554-1104
Supplier:  VICI JOUR RESEARCH
undefined
Guillotine tubing cutter
Catalog # 554-1104
Supplier:  VICI JOUR RESEARCH
Specifications
  • Description:
    Guillotine tubing cutter
  • Pk:
    1
Specifications
About this item
Cutter for cutting PTFE, PEEK and other polymeric tubing.