Tubing, Tygon® 3350
Silicone, platinum cured, translucent.
- Biocompatible, ideal for sensitive applications, can be used for foodstuffs
- Resistant to ozone and UV, autoclavable
Tygon® tubing made from platinum-cured silicone with ultra-smooth surface. High elasticity with good restoring force for constant liquid transfer, even with intermittent pumping action.
Temperature resistance: −80 to +204 °C
Hardness (Shore durometer A): 50
Density: 1,14
: FDA approved, compliant with USP class VI.
: Tubing is double bagged inside a cardboard box. Labelling is on the inner bag and the box with certificate of conformity and date of manufacture.