Air bleeds
4 mm, 180 mm
201-2610
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Hollow prevents boiling splashes.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Ø:4 mm
- Length:180 mm