Test tubes with rim
Supplier: SciLabware
Jump to:
About this item
PYREX®, borosilicate glass, heavy wall.
- Excellent chemical resistance
- High temperature resistance
Heavy wall to provide extra mechanical strength. Robust construction reduces possibility of breakages. Improved user safety.
Certifications: ISO 4142
4 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, heavy wall.
- Excellent chemical resistance
- High temperature resistance
Heavy wall to provide extra mechanical strength. Robust construction reduces possibility of breakages. Improved user safety.
Certifications: ISO 4142
Product Family Options
Product Information