PCR microtube strips, 0.2 ml, Amplitube™
PP. Ultra-thin wall design ensures uniform and efficient thermal transfer, therefore reducing PCR reaction time in most V-bottomed thermal cyclers.
- Will fit standard well spacing and can be used with 8-channel pipettes
- Strips can be cut and used as individual tubes
- Choice of natural and four non-cytotoxic and non-metallic colours for visual coding of samples
- Non-attached, domed cap strips, which must be ordered separately, ensure closure during the whole thermal cycle
- Certified RNase, DNase and DNA-free
Ultra clear, flat caps are ideal for fluorescence, luminescence and real-time PCR applications.
: In tamper-proof, re-sealable bags.