Glass ceramic laboratory protection plates, DURAN®
These quadrupods are made from heat-resistant chrome-nickel steel with four legs for extra stability.
- Uniformity of heating distribution
- Excellent energy transfer
- Plates can either be used with quadrupods or with plate holders
Temperature: −200 to +700 °C