ContactUsLinkComponent
Beaker tong, steel coated
Beaker tong, steel coated
Catalog # 231-4120
Supplier:  BOCHEM
undefined
Beaker tong, steel coated
Catalog # 231-4120
Supplier:  BOCHEM
Specifications
  • Length:
    300 mm
  • Pk:
    1
  • Spanning area:
    32 - 140 mm
Specifications
About this item
Steel, coated. For secure, easy handling of beakers and similar containers.
The jaws of the tongs are equipped with special silicone cushioning, which not only protect the containers from being damaged by the jaws, but also provide thermal insulation.