Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Amarell
Precision thermometers
Supplier:  Amarell
Certificates
Precision thermometers
Precision thermometers
2 Options Available Below
Product Details & Documents
These total immersion thermometers with enclosed scale are suitable for government verification. Red spirit.
  • Safe and environmentally friendly
  • Milk glass scale
  • Colourless capillary
Documents
Product Family Options
With calibration certificate
Product Information
  • Range
    Accuracy
    Length
    Ø
    Availability
    Price
  • 0…+100 °C
    ±0,1 %
    550 mm
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
With DKD certificate
Product Information
  • Range
    Accuracy
    Length
    Ø
    Availability
    Price
  • 0...+100 °C
    ±0,1 %
    550 mm
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST