Orders LinkContactUsLinkComponent
pH indicator papers, MQuant®, Supelco®
 :   Merck
pH indicator papers, MQuant®, Supelco®
 :  Merck
pH indicator papers, MQuant®, Supelco®
pH indicator papers, MQuant®, Supelco®
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
  • Catalog No:
  • 1.09527.0001
  • 1.10232.0001
  • 1.09526.0003
  • 1.09486.0003
  • 1.10962.0003
  • 1.09489.0003
  • 1.09521.0003
  • 1.09555.0003
  • 1.09557.0003
  • 1.09514.0003
  • 1.09556.0003
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
Product Family Options
  • Description
    pH range
    pH gradation
  • Universal indicator replacement rolls only*
    1 - 10
    1
    1 * 6 Rolle
    1 * 6 Rolle - 60,10 €
    60,10 €
    1 * 6 Rolle
       60,10 €
    Specifications
    Cat. No.
    1.09527.0001
    M-Clarity quality level
    MQ100
  • Replacement rolls for pH 1 - 14*
    -
    1
    1 * 6 Rolle
    1 * 6 Rolle - 58,90 €
    58,90 €
    1 * 6 Rolle
       58,90 €
    Specifications
    Cat. No.
    1.10232.0001
    M-Clarity quality level
    MQ100
  • Universal indicator, including colour scale
    1 - 10
    1
    1 * 3 Rolle
    1 * 3 Rolle - 44,80 €
    44,80 €
    1 * 3 Rolle
       44,80 €
    Specifications
    Cat. No.
    1.09526.0003
    M-Clarity quality level
    MQ100
  • Litmus paper blue (pH<7 red / >7 blue)
    -
    -
    1 * 3 Rolle
    1 * 3 Rolle - 45,70 €
    45,70 €
    1 * 3 Rolle
       45,70 €
    Specifications
    Cat. No.
    1.09486.0003
    M-Clarity quality level
    MQ100
  • Universal indicator
    1 - 14
    1
    1 * 3 Rolle
    1 * 3 Rolle - 46,40 €
    46,40 €
    1 * 3 Rolle
       46,40 €
    Specifications
    Cat. No.
    1.10962.0003
    M-Clarity quality level
    MQ100
  • Litmus paper red (pH<7 red / >7 blue)
    -
    -
    1 * 3 Rolle
    1 * 3 Rolle - 46,50 €
    46,50 €
    1 * 3 Rolle
       46,50 €
    Specifications
    Cat. No.
    1.09489.0003
    M-Clarity quality level
    MQ100
  • Phenolphthalein paper (pH<8,5 colourless / >8,5 red)
    -
    -
    1 * 3 Rolle
    1 * 3 Rolle - 55,70 €
    55,70 €
    1 * 3 Rolle
       55,70 €
    Specifications
    Cat. No.
    1.09521.0003
    M-Clarity quality level
    MQ100
  • Special indicator
    3,8 - 5,4
    0,2 - 0,3
    1 * 3 Rolle
    1 * 3 Rolle - 53,60 €
    53,60 €
    1 * 3 Rolle
       53,60 €
    Specifications
    Cat. No.
    1.09555.0003
    M-Clarity quality level
    MQ100
  • Special indicator
    6,4 - 8,0
    0,2 - 0,3
    1 * 3 Rolle
    1 * 3 Rolle - 53,60 €
    53,60 €
    1 * 3 Rolle
       53,60 €
    Specifications
    Cat. No.
    1.09557.0003
    M-Clarity quality level
    MQ100
  • pH-Indicator paper congo red
    -
    -
    1 * 3 Rolle
    1 * 3 Rolle - 50,20 €
    50,20 €
    1 * 3 Rolle
       50,20 €
    Specifications
    Cat. No.
    1.09514.0003
    M-Clarity quality level
    MQ100
  • Special indicator
    5,4 - 7,0
    0,2 - 0,4
    1 * 3 Rolle
    1 * 3 Rolle - 51,30 €
    51,30 €
    1 * 3 Rolle
       51,30 €
    Specifications
    Cat. No.
    1.09556.0003
    M-Clarity quality level
    MQ100