Aquarium tanks
Supplier: MARIENFELD
New Product
Certificates
About this item
Aquarium tanks for a large range of applications. Made from clear soda glass.
- Strength, thick-walled
- Ground rim
- For applications without large demands on temperature stability