Special hypodermic needles, long, Vet Premium
Catalog # 613-3906
Supplier: Air-Tite Products
Specifications
- Gauge:18
- Description:Vet Premium hypodermic needles
- Length:3''
- Seq:05
- Pk:100
Specifications
About this item
Stainless steel needles conform to the strictest quality control standards. For veterinary use and laboratory use only.
- Precise bevelled point
- For Luer tips
Packaging: Individually packed.