Syringes, medical
Catalog #
Product Details & Documents
Sterile, graduated, single use, three pieces, body made of transparent PP, white plunger in PP/titanium oxide and with silicone seal, no latex.
- Standard Luer Tip (LT) or screw connector Luer Lock (LL), centred (1 to 3 ml) or off-centred (5 to 100 ml)
- The watertight silicone seal reduces contact with the solution and guarantees minimal risk of particulate contamination of the solution
- Sterilised with ethylene oxide (EN 550)
: Conforms to ISO 9001, EN 46001, medical guideline 93/42/EEC
: In individual blister packs.