VWR® M40, Transport Swab
Catalog # 710-0442
Supplier: VWR Collection
Specifications
- Description:Transport swabs, M40
- Material:Plastic body
- Transport medium:Amies gel
- Pk:50
Specifications
About this item
Transport swab with plastic shaft, synthetic tip, and Amies agar gel.
- Compliant with CLSI (formerly NCCLS) standard M40-A at 21 and 4 °C
- Without charcoal
Helps to improve recovery and survival of fastidious microorganisms from the throat, vagina, wounds and skin swabs.
Certifications: CLSI (formerly NCCLS).
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904