Bench protection papers
These protection papers are available in different formats. They can be used for bench protection as well as for the packaging of glass parts.
- Sheets are available flat or folded
- Suitable for food contact
- Cellulose and cotton
Weight: 65 g/m²
Porosity: 7 to 10 µm
Weight: 65 g/m²
Thickness: 0,16 mm
Porosity: 7 - 10 µ