Orders LinkContactUsLinkComponent
Stoppers, rubber, conical, without holes
Stoppers, rubber, conical, without holes
Catalog #
Stoppers, rubber, conical, without holes
Catalog #
Supplier:  
Stoppers, rubber, conical, without holes
Stoppers, rubber, conical, without holes
Product Details & Documents
Conical natural grey-blue rubber stoppers without holes.
  • Resistant to common laboratory chemicals
  • Highly elastic, for a perfect seal and exceptional bonding
  • Solid form
  • Resistant to temperatures from −5 to +70 °C (+120 °C in humid atmosphere)
Resistant to temperatures from -5 to +70 °C (120 °C in humid atmosphere)
DIN 12871
Certifications: DIN 12871