VWR®, Magnetic Stirrers, with Crosshead
Supplier: VWR Collection
Certificates
About this item
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly suitable for small stirring vessels, extremely effective stirring action.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)