VWR®, Magnetic stirring bars, oval
Supplier: VWR Collection
Certificates
About this item
PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)