Electronic diamond scriber, EDS 20
Supplier: MEDITE
Certificates
About this item
For inscribing object holders and marking, inscribing or etching metal or plastic. Manually bringing the scribe into the writing position will automatically activate the motor; when the scribe is returned to a horizontal position, the motor switches itself off. When in the writing position, the tip rotates very quickly, thereby enabling effortless etching. Weight approx. 100 g, average finger grip Ø ~15 mm.
- Tips are light and can be replaced without the need for tools
- Inscription is etched so deep that it can last almost indefinitely