Wet surface pen, SP Bel-Art
Catalog # 817-1135
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Wet surface pen
- Pk:1
Specifications
About this item
It can be used to correct or write on wet labels that were kept in a water bath, in liquid nitrogen or in a freezer at 0 °C. Can be also used on dry paper as a standard ball-point pen.
- Writes clearly on wet surfaces