Chemical spoon with spatula end
Supplier: RSG
Jump to:
About this item
Stainless steel Remanit 4301, anti-magnetic. Multipurpose spoon for a wide range of uses.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
6 Options Available Below
Product Details & Documents
Stainless steel Remanit 4301, anti-magnetic. Multipurpose spoon for a wide range of uses.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Product Family Options
Product Information
- LengthSpoon sizeAvailabilityPrice