Orders LinkContactUsLinkComponent
Spoons, POLY
Spoons, POLY
Supplier:   BOCHEM
Spoons, POLY
Supplier:  BOCHEM
Certificates
Spoons, POLY
11 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Spoon size
    Availability
    Price
  • 231-1068
    Straight
    Straight
    155 mm
    40×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,00 €
    5,00 €
    1 * 1 ST
       5,00 €
  • 231-1069
    Straight
    Straight
    185 mm
    40×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,60 €
    5,60 €
    1 * 1 ST
       5,60 €
  • 231-1261
    Sharply oblique
    Sharply oblique
    190 mm
    25×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,50 €
    4,50 €
    1 * 1 ST
       4,50 €
  • 231-1070
    Straight
    Straight
    210 mm
    40×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,00 €
    6,00 €
    1 * 1 ST
       6,00 €
  • 231-1296
    Oblique
    Oblique
    215 mm
    70×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,60 €
    7,60 €
    1 * 1 ST
       7,60 €
  • 231-0604
    Oblique
    Oblique
    250 mm
    65×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,80 €
    8,80 €
    1 * 1 ST
       8,80 €
  • 231-1269
    Straight
    Straight
    255 mm
    40×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,60 €
    6,60 €
    1 * 1 ST
       6,60 €
  • 231-1071
    Straight
    Straight
    300 mm
    40×20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,00 €
    7,00 €
    1 * 1 ST
       7,00 €
  • 231-1297
    Oblique
    Oblique
    300 mm
    65×28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,80 €
    9,80 €
    1 * 1 ST
       9,80 €
  • 231-1270
    Oblique
    Oblique
    395 mm
    65×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,90 €
    12,90 €
    1 * 1 ST
       12,90 €
  • 231-0605
    Oblique
    Oblique
    500 mm
    65×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,30 €
    14,30 €
    1 * 1 ST
       14,30 €