Pharmacist's spoon
150 mm
Catalog # 231-1387
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses.
- Narrow shape with extra long, flat handle
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Documents
RSGA028.150_TDS_EN
Datasheet
Specifications
- Length:150 mm