Order Entry
Germany
Orders LinkContactUsLinkComponent
Laboratory spoon

Laboratory spoon

200 mm, 30×16 mm
Catalog # 231-1231
Supplier:   RSG
Laboratory spoon
Catalog # 231-1231
Supplier:  RSG
Laboratory spoon
Laboratory spoon

200 mm, 30×16 mm

Price
12,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses.

  • Open ended spoon, with flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    200 mm
  • Spoon size:
    30×16 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.