Orders LinkContactUsLinkComponent
Double spatulas
Double spatulas
Supplier:   BOCHEM
Double spatulas
Supplier:  BOCHEM
Certificates
Double spatulas
14 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Availability
    Price
  • 130 mm
    40 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,00 €
    2,00 €
    1 * 1 ST
       2,00 €
  • 130 mm
    40 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 9,90 €
    9,90 €
    1 * 5 ST
       9,90 €
  • 150 mm
    45 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,30 €
    2,30 €
    1 * 1 ST
       2,30 €
  • 150 mm
    45 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 11,20 €
    11,20 €
    1 * 5 ST
       11,20 €
  • 185 mm
    50 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,40 €
    2,40 €
    1 * 1 ST
       2,40 €
  • 185 mm
    50 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 12,00 €
    12,00 €
    1 * 5 ST
       12,00 €
  • 210 mm
    60 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,60 €
    2,60 €
    1 * 1 ST
       2,60 €
  • 210 mm
    60 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 19,70 €
    19,70 €
    1 * 5 ST
       19,70 €
  • 250 mm
    70 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,10 €
    3,10 €
    1 * 1 ST
       3,10 €
  • 250 mm
    70 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 15,50 €
    15,50 €
    1 * 5 ST
       15,50 €
  • 300 mm
    75 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,00 €
    4,00 €
    1 * 1 ST
       4,00 €
  • 300 mm
    75 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 20,00 €
    20,00 €
    1 * 5 ST
       20,00 €
  • 400 mm
    100 mm
    16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,70 €
    7,70 €
    1 * 1 ST
       7,70 €
  • 500 mm
    100 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,60 €
    9,60 €
    1 * 1 ST
       9,60 €