Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spatulas
Spoon spatulas
Spoon spatulas
Spoon spatulas
Supplier:
MORGAN ADVANCED MATERIALS HALD
Certificates
Expand image
Spoon spatulas
Supplier:
MORGAN ADVANCED MATERIALS HALD
About this item
Porcelain, glazed white.
Broad shape
Excellent chemical resistance
Heat resistant up to 1000 °C
Designed for rough handling