ContactUsLinkComponent
Dispenser for Parafilm® M sealing film
Dispenser for Parafilm® M sealing film
Catalog # 291-1219
Supplier:  Brand
undefined
Dispenser for Parafilm® M sealing film
Catalog # 291-1219
Supplier:  Brand
Specifications
  • Description:
    Dispenser with safety cutter for Parafilm® M
  • Pk:
    1
Specifications
About this item
In plastic, light grey. For neat and safe cutting of Parafilm® M sealing film as well as clean storage.
  • For rolls 50 or 100 mm in width
  • Film feed device
  • Safety cutter