Adhesive PCR film and foil seals
Supplier: Thermo Fisher Scientific
Certificates
About this item
Clear polyester film, aluminium foil. Recommended for use in PCR in conjunction with a thermal cycler that has a screw-down or clip-down lid.
- Strong adhesive prevents evaporation during thermal cycling or sample storage
- Transparent film for fluorescence applications
- No sticky residue on the plate when seal is removed
- Temperature range: for film −20 to +120 °C; for foil −80 to +150 °C