Adhesive PCR sealing film and foil seals
Transparent adhesive sealing film is ideal for reducing cross contamination and/or spill over in PCR applications.
- Thermostable and functional from −70 to +95 °C
- Choice of sterile or non sterile
Aluminium sealing foil is suitable for both manual sealing during PCR work and high throughput applications. Pierceable with a pipette tip for easy access to sample. Adhesive backing makes it easy to apply. Will resist temperatures from –80 to +120 °C. It is recommended that the foil be applied using the specially designed rubber roller to ensure a perfect bond, eliminating the risk of evaporation.