Measuring scoops
Supplier: Brand
Jump to:
About this item
PP, white. With flat, bevelled end and reinforced handle.
- Excellent chemical resistance, unbreakable
- The capacity of the scoop is stamped into the handle for easy reading
5 Options Available Below
Product Details & Documents
PP, white. With flat, bevelled end and reinforced handle.
- Excellent chemical resistance, unbreakable
- The capacity of the scoop is stamped into the handle for easy reading