Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
Supplier:   VITLAB
Measuring scoops
Supplier:  VITLAB
Certificates
Measuring scoops
Measuring scoops
9 Options Available Below
Product Details & Documents
PP, white. With flat, chamfered ends and reinforced handle.
  • Excellent chemical resistance, robust
  • Heat resistant up to +130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 2 ml
    60 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 5,10 €
    5,10 €
    1 * 12 ST
       5,10 €
  • 5 ml
    82 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 5,80 €
    5,80 €
    1 * 12 ST
       5,80 €
  • 10 ml
    100 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 7,20 €
    7,20 €
    1 * 12 ST
       7,20 €
  • 25 ml
    135 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 9,50 €
    9,50 €
    1 * 12 ST
       9,50 €
  • 50 ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 13,80 €
    13,80 €
    1 * 12 ST
       13,80 €
  • 100 ml
    200 mm
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 20,60 €
    20,60 €
    1 * 12 ST
       20,60 €
  • 250 ml
    260 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 22,70 €
    22,70 €
    1 * 6 ST
       22,70 €
  • 500 ml
    315 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 32,00 €
    32,00 €
    1 * 6 ST
       32,00 €
  • 1000 ml
    385 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 38,50 €
    38,50 €
    1 * 6 ST
       38,50 €